Dustless Precision Pellets® Primate Purified Diet, Bio-Serv
Supplier: Bio-Serv
F0079 F0035 F0022 F0059 F0059
CA89067-532EA 724.68 CAD
CA89067-532 CA89067-536 CA89067-544 89067-534 CA89067-534
Dustless Precision Pellets® Primate Purified Diet, Bio-Serv
Animal Supplies Animal Feed
Precise size and weight pellets generally used in nutritional and behavioral studies.
  • Dustless and fracture free
  • Superior performance in automatic feeders
  • Nutritionally complete, banana-flavored, grain-based formula
  • Available in purified formula
  • Wide range of weights: 20 mg to 1 g
  • Every batch tested for hardness, weight and height
  • Custom flavors and colors available upon request
  • Nutritionally assayed
  • Shelf Life: 12 months, cool dry storage
Ingredients: Casein, Dextrose, Sucrose, Cellulose, Corn Starch, Corm Oil, Corn Syrup, Mineral Mix, CholineMore Product Information