Dustless Precision Pellets® Primate Grain-Based Diet, Bio-Serv

Supplier: Bio-Serv
F0158 F0179 F0157 F0157
CA89067-520EA 798.86 CAD
CA89067-520 CA89067-522 CA89067-524 CA89067-530 89067-530
Dustless Precision Pellets® Primate Grain-Based Diet, Bio-Serv
Animal Supplies Animal Feed
Precise size and weight pellets generally used in nutritional and behavioral studies.

  • Dustless and fracture free
  • Superior performance in automatic feeders
  • Nutritionally complete, banana-flavored, grain-based formula
  • Available in purified formula
  • Wide range of weights: 20 mg to 1 g
  • Every batch tested for hardness, weight and height
  • Custom flavors and colors available upon request
  • Nutritionally assayed
  • Shelf Life: 12 months, cool dry storage

Ingredients: Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (preserved with BHA), Dried Brewers Yeast, Mineral Mix, Banana Flavor, Vitamin Mix, Magnesium Stearate, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin.

Caloric value: 3.35 kcal/g
Nutritional ash content: 7.8%
Nutritional carbohydrate content: 54.0%
Nutritional fat content: 3.8%
Nutritional fibre content: 4.0%
Nutritional moisture content: <10%
Nutritional protein content: 21.3%
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR