Primate Medications, Bio-Serv
Supplier: Bio-Serv
MD050-50 MD100-80 MD200-120 MD750-100 MD800-100 MD950-4 MD150-15 MD975-40 MD350-20 MD300-20 MD600-100 MD500-50 MD900-222 MD800-100 MD750-250 MD925-500 F05235 F05237 MD700-100 MD275-080 MD275-080
CA89067-484PK 865.41 CAD
CA89067-484 CA89067-486 CA89067-488 CA89067-490 CA89067-492 CA89067-498 CA89067-500 CA89067-502 CA89067-504 CA89067-506 CA89067-508 CA89067-510 CA89067-512 CA89067-496 89067-492 CA89408-210 CA89408-212 CA89408-214 CA89408-216 CA89408-218 76353-780 CA76353-780
Primate Medications, Bio-Serv
Animal Supplies Animal Feed
Chewable, flavored, medicated 5 g tablet replaces more stressful dosing methods
Base Meal Ingredients: Sucrose, Dextrose, Mineral Oil, Artificial and Natural Flavors, Calcium Silicate, Magnesium Stearate.
Note: A signed MD Form is required for all Medications except Aspirin and L-Tryptophan. Please Fax the completed form to 908-284-4753. The form will remain on file for one year. Refills are permitted as long as the prescribing veterinarian listed on the request form is still affiliated with the requesting institution.
Tablet consistency ensures accurate dosing; scored for flexible dose administration. Cold tableting process eliminates drug degradation. No heat or water used in processing. Highly palatable sugar-based formula with excellent drug-masking capabilities. Reduces time, labor and stress compared to traditional dosing methods. Medications can be customized to meet specific requirements. Shelf Life: 6-24 months as dictated by the drug expiration date.