A 10 x's lower dosage that can be successfully used in breeding animals with out negatively affecting reproduction. An effective method to reduce the hazards of respiratory problems in immunocompromised mice.
- Highly palatable, nutritionally complete, grain-based, bacon-flavored tablets
- More convenient and minimizes risk of dehydration compared to using medicated water
- Easy to administer; place on cage floor for easy access
- Direct compression process eliminates drug degradation
- Sterilized by gamma irradiation
- Nutritionally assayed
- Shelf Life: 12 months, cool dry storage
Ingredients: Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (Preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Magnesium Stearate, Calcium Silicate, Bacon Flavor, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin, Sulfamethoxazole, Trimethoprim.
Each 5 g bacon-flavored scored tablet contains 6 mg Sulfamethoxazole and 1.2 mg Trimethoprim. Mouse average dose is one 5 g tablet per 20 g of body weight.
Caloric value: 3.39 kcal/g
Nutritional ash content: 7.8%
Nutritional carbohydrate content: 54.0%
Nutritional fat content: 4.3%
Nutritional fibre content: 4.0%
Nutritional moisture content: <10%
Nutritional protein content: 21.3%