Rodent Foraging Crumbles, Certified, Bio-Serv

Supplier: Bio-Serv
F5783 S5783 S5783 F5783
76406-496BG 84.74 CAD
76406-496 CA71002-336 89408-276 CA76406-496
Rodent Foraging Crumbles, Certified, Bio-Serv
Animal Supplies Animal Feed
Nutritionally complete, grain-based treat

  • The tiny crumbles encourage foraging behavior
  • Scatter a pinch directly into bedding
  • Bacon flavor enhances foraging activity
  • Nutritionally assayed

Ingredients: Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (Preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Bacon Flavor, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin.

Caloric value: 3.41 kcal/g
Nutritional ash content: 7.4%
Nutritional carbohydrate content: 54%
Nutritional fat content: 4.3%
Nutritional fibre content: 4%
Nutritional moisture content: <10%
Nutritional protein content: 21.6%

Certifications: Certified (Contaminant Screened).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR