Rodent Diet, AIN-93M, Bio-Serv

Supplier: Bio-Serv
F3155 F3198
CA89408-268EA 5476.09 CAD
CA89408-268 CA89408-270
Rodent Diet, AIN-93M, Bio-Serv
Animal Supplies Animal Feed
A purified diet formulated to meet the nutritional requirements of mature rodents.

  • Widely used and published
  • Can be customized to meet research objectives
  • Phytoestrogen free
  • Chlorophyll free; does not interfere with imaging studies
  • Nutritionally assayed
  • Shelf Life: 6 months refrigerated

Ingredients: Corn Starch, Casein, Maltodextrin, Sucrose, Cellulose, Soybean Oil, Mineral Mix, Vitamin Mix, Choline Bitartrate, L-Cystine, tBHQ.

Caloric value: 3.58 kcal/g
Nutritional ash content: 2.2%
Nutritional carbohydrate content: 67.5%
Nutritional fat content: 4.1%More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR