B-Wholesome Treats™, Certified, Bio-Serv

Supplier: Bio-Serv
F7500 F10507 F10507
CA89408-186EA 433.22 CAD
CA89408-186 77391-938 CA77391-938
B-Wholesome Treats™, Certified, Bio-Serv
Animal Supplies Animal Feed
Whole wheat-based treat made with 100% USDA organic ingredients.

  • Healthy alternative to the average biscuit
  • Nutritionally assayed
  • Shelf Life: 12 months, cool dry storage

Ingredients: Organic rye flour, blueberries, organic olive oil, organic eggs, organic nonfat dry milk, organic honey, organic garlic.

Caloric value: 3.95 kcal/g
Nutritional ash content: 1.6%
Nutritional carbohydrate content: 72.2%
Nutritional fat content: 7.5%
Nutritional fibre content: 1.7%
Nutritional moisture content: <10%
Nutritional protein content: 9.5%

Certifications: Certified (Contaminant Screened).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR