Fruit Crunchies™ Treats, Certified, Bio-Serv
Supplier: Bio-Serv
F05798 S05798-1 F05798 S05798-1 F05798-1
CA89067-430EA 707.37 CAD
CA89067-430 CA89067-432 89067-430 89067-432 CA11024-216
Fruit Crunchies™ Treats, Certified, Bio-Serv
Animal Supplies Animal Feed
Nutritionally complete 190 mg treat that complements enrichment programs can be used as a reward or foraging treat.
  • A medley of apple, grape and orange flavors adds variety to foraging activity
  • Excellent source of Vitamin C
  • Less labor intensive and more sanitary than perishable fruit
  • Sterile version is sterilized by gamma irradiation
  • Nutritionally assayed
  • Shelf Life: 12 months, cool dry storage
Ingredients: Casein, Dextrose, Sucrose, Cellulose, Corn Starch, Corn Oil, Corn Syrup, Mineral Mix, Choline Bitartrate, Vitamin Mix, Natural and ArtificialMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable