Bacon Softies™ Treats, Certified, Bio-Serv
Supplier: Bio-Serv
F3580 S3580-1 F3580 S3580-1 F3580-1
CA89067-424EA 500.24 CAD
CA89067-424 CA89067-426 89067-424 89067-426 CA76353-750
Bacon Softies™ Treats, Certified, Bio-Serv
Animal Supplies Animal Feed
Soft, pelleted, nutritionally complete diet for rodents and supplement for canines and ferrets.
  • Highly palatable bacon flavor stimulates appetite
  • Excellent soft diet for recovering or debilitated animals and weanlings
  • Replaces labor intensive, nutrient-diluted mush diets
  • Available sterile
  • Nutritionally assayed
  • Shelf Life: 6 months, refrigerated
Ingredients: Soy Flour, Soybean Oil, Corn Syrup, Sucrose, Casein, Cellulose, Corn Starch, Wheat Bran, Molasses, Mineral Mix, Banana Flakes, Acacia Gum, Bacon Flavor, Glycerin, Vitamin Mix, DL-Methionine,More Product Information