Extran® MA 02 Liquid Detergent for Manual Washing, MilliporeSigma
Supplier: MilliporeSigma
1.07553.5000 1.07553.2500 1.07553.9010 1.07553.9025
CA1.07553.5000CS 1161 CAD
CA1.07553.5000 CA1.07553.2500 CA1.07553.9010 CA1.07553.9025
Extran® MA 02 Liquid Detergent for Manual Washing, MilliporeSigma
Cleaning Agents Detergents and Surface Cleansers
Universal cleaner for the gentle cleaning of appliances made from alkali-sensitive metals such as aluminum, zinc and alloys with similar behavior. Suitable for metal appliances and precision measuring devices made from glass and quartz such as burettes, pipettes, cells, blood gas analyzers and other medical equipment which is sensitive to aggressive detergents and also rarely has problematic contamination.
  • Liquid
  • Neutral
  • Chlorine-free
  • Free from odorants/dyestuffs
Dosing: The ideal dosing depends on the hardness of the water and the level ofMore Product Information