Extran® MA 01 Liquid Detergent for Manual Washing, MilliporeSigma

Supplier: MilliporeSigma
EX0993-20 1.07555.5000 1.07555.1000 1.07555.2500 1.07555.9010 1.07555.9025
CAEX0993-20EA 2811 CAD
CAEX0993-20 CA1.07555.5000 CA1.07555.1000 CA1.07555.2500 CA1.07555.9010 CA1.07555.9025
Extran® MA 01 Liquid Detergent for Manual Washing, MilliporeSigma
Cleaning Agents Detergents and Surface Cleansers
Extran® MA 01 is a universal cleaner for the removal of heavy contamination. In wiping tables, tiles, floors. In soaking for the automated cleaning of laboratory equipment.

  • Liquid
  • Alkaline
  • Chlorine-free
  • Free from odorants/dyestuffs

Dosing
The ideal dosage depends on the hardness of the water and the level of contamination of the item to be cleaned.

Recommended application concentrations:
- For normal contamination: 2%
- For heavier contamination: 5%
- For very tough stains up to 20%

pH value (5%; water) 11.0–12.0

CAS: 103171-11-5

Order Now

SPECIFICATIONS

Environmentally Preferable
Formulation Ionic and non-ionic surfactants, phosphates, excipients in low quantities
pH 11.6 (20 g/L, H₂O, 20 °C)
Boiling point 100 °C (1013 hPa)
Solubility (20 °C) Soluble
Density 1.07 g/cm³ (20 °C)
Vapor pressure 23.4 hPa (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR