Extran® AP 21 Acid Rinse with Phosphoric Acid for Automatic Washing, MilliporeSigma
Supplier: MilliporeSigma
1.07559.2500 1.07559.9010 1.07559.9025
CA1.07559.2500EA 130 CAD
CA1.07559.2500 CA1.07559.9010 CA1.07559.9025
Extran® AP 21 Acid Rinse with Phosphoric Acid for Automatic Washing, MilliporeSigma
Cleaning Agents Detergents and Surface Cleansers
Extran® AP 21 The acid special cleaner can be used both as a prewash agent and a rinsing agent with a neutralising effect. When used as a pre-wash agent, it primarily dissolves carbonates and hydroxides from the residues. Protein substances and organic bases, such as amines, are often removed better in an acidic prewash as in an alkaline main wash cycle. As a rinsing agent, i.e. after the alkaline main wash cycle, it is especially suitable for removing remaining traces of alkali on the cleaned material or, in the case of solution carry-over, for neutralisation. This acidic cleaning agent isMore Product Information