NEO Micro Cover Glass, No.1, Rectangular, Matsunami Glass
Supplier: Matsunami Glass
C024501 C024601 C024401
10748-188BX 85.37 CAD
10748-188 10748-190 10748-187
NEO Micro Cover Glass, No.1, Rectangular, Matsunami Glass
Cover Glasses
Matsunami Glass is a pioneer in the development of cover glass, and widely recognized as the standard for quality and flatness
When working with an automated coverslipper a great deal of stress is applied to the cover glass by the feeding mechanism. NEO Micro Cover Glass can withstand twice the bending stress of competitor products.
Some cover glass tend to stick together in hot, humid conditions. NEO Micro Cover Glass has been specially treated with a coating to prevent sticking. This coating does not interfere with optical transmission.
Mounting media spreads evenly and remainsMore Product Information