Rhesus Liquid Primate Diet, Control, Bio-Serv

Supplier: Bio-Serv
F0550SP F0550SP
CA89067-388EA 325.91 CAD
CA89067-388 89067-388
Rhesus Liquid Primate Diet, Control, Bio-Serv
Animal Supplies Animal Feed
Highly palatable, banana-flavored nutritional diet

  • Recommended for research studies and special needs
  • Custom formulations available
  • Medications can be added
  • Easy to prepare; no blender needed. Add water, shake and pour. Use with Bio-Serv's 160 oz. Shake and Pour Bottle or your own mixing vessel
  • Nutritionally assayed
  • Shelf Life: Dry Mix, 6 months refrigerated; Prepared, 3 days

Ingredients: Corn Syrup Solids, Casein, Maltodextrin, Olive Oil, Cellulose, Mineral Mix, Corn Oil, Vitamin Mix, Suspending Aids, Banana Flavor, Choline Chloride, L-Cystine, DL- Methionine.

Caloric value: 1010 kcal/L
Nutritional ash content: 3.7%
Nutritional carbohydrate content: 52.0%
Nutritional fat content: 9.3%
Nutritional fibre content: 4.7%
Nutritional moisture content: <10%
Nutritional protein content: 22.9%
Vitamin C: 0.6 g/L
Vitamin D3: 2000 IU/L
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR