PRIMA-Treats®, Bio-Serv

Supplier: Bio-Serv
F0295 F0300 F0296 F0309 F0308 F05709 F0345 F0407 F0408 F0482 F0295 F0300 F0296 F0309 F0308 F05709 F0345 F0407 F0408 F0293 F0293
CA89067-352PK 511.15 CAD
CA89067-352 CA89067-354 CA89067-356 CA89067-358 CA89067-362 CA89067-366 CA89067-368 CA89067-376 CA89067-378 CA89067-380 89067-352 89067-354 89067-356 89067-358 89067-362 89067-366 89067-368 89067-376 89067-378 76419-520 CA76419-520
PRIMA-Treats®, Bio-Serv
Animal Supplies Animal Feed
Nutritionally complete treat that complements enrichment programs

  • Excellent source of Vitamin C
  • Use for training or positive reward following a less desirable procedure
  • Less labor intensive and more sanitary than perishable fruit
  • Uncolored Orange and Assorted Fruit flavors eliminate color bias, are Certified (Contaminant Screened), and suitable for use in GLP and TOX studies
  • Nutritionally assayed
  • Shelf Life: 12 months, cool dry storage

Ingredients: Dextrose, Casein, Sucrose, Cellulose, Mineral Mix, Canola Oil, Natural and Artificial Flavors, Magnesium Silicate, Choline Dihydrogen Citrate, Vitamin Mix, Calcium Silicate, Ascorbic Acid, DL-Methionine, Magnesium Stearate, Food Dye (Excluding Uncolored Flavors), BHA.

Caloric value: 16.4 kcal/Wafer
Nutritional ash content: 6.3%
Nutritional carbohydrate content: 52.5%
Nutritional fat content: 5.0%
Nutritional fibre content: 11.2%
Nutritional moisture content: <10%
Nutritional protein content: 18.1%
Vitamin C: 21.9 mg/Wafer
Vitamin D3: 10.1 IU/Wafer
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR