PRANG™ Oral Rehydrator, Certified, Bio-Serv

Supplier: Bio-Serv
F2351-A F2351-C F2351-O F2351-B F2351-G F2351-S F2351-A-GAL F2351-B-GAL F2351-C-GAL F2351-G-GAL F2351-O-GAL F2351-S-GAL
CA89067-326EA 425.11 CAD
CA89067-326 CA89067-328 CA89067-330 CA89067-332 CA89067-334 CA89067-336 CA89408-240 CA89408-242 CA89408-244 CA89408-246 CA89408-248 CA89408-250
PRANG™ Oral Rehydrator, Certified, Bio-Serv
Animal Supplies Animal Feed
Flavored oral rehydrator with electrolytes and Vitamin C in a powder form.

  • Excellent for crashing, recovering and debilitating conditions
  • Balanced mineral and glucose levels for quick energy
  • Combines the benefits of both Gatorade and Tang with less sugar
  • Nutritionally assayed
  • Just add water - 0.1 kg/Jar makes 1 gallon; 1 kg/Jar makes 10 gallons
  • Shelf Life: Dry Mix, 12 months, cool dry storage; Prepared, 7 days refrigerated

Ingredients: Dextrose, Sodium Chloride, Sodium Bicarbonate, Citric Acid, Potassium Chloride, Ascorbic Acid, Acesulfame Potassium, Natural and Artificial Flavors, Food Dye.

Nutritional carbohydrate content: kcals/8 oz. 19.88%

Certifications: Certified (Contaminant Screened).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR