Wood Enrichment Devices, Bio-Serv
Supplier: Bio-Serv
W0003 W0005 W0005 W0003 W0026
CA89067-800EA 29.44 CAD
89067-800 CA89067-800 89067-806 CA89067-806 76285-300
Wood Enrichment Devices, Bio-Serv
Animal Supplies Animal Environmental Enrichment
Made of natural, non-injurious hardwood
Promotes species-typical behavior and counteracts boredom. Benefits dental health. Hide treats in the two 3/4" diameter pre-drilled holes. Autoclavable.