Nalgene® Syringe Filters, PTFE, 25 mm, Thermo Scientific

Supplier: Thermo Fisher Scientific
728-2020 728-2045
10218-498CS 4906.84 CAD
10218-498 10218-500
Nalgene® Syringe Filters, PTFE, 25 mm, Thermo Scientific
Filters Syringe Filters
Filter solvents with Thermo Scientific™ Nalgene™ 25 mm Ø syringe filters with PTFE membranes.

  • Ideal for chromatography sample preparation
  • Accommodate sample volumes from 10 to 100 ml
  • White housing ring
  • Non sterile, Autoclavable

For use with aggressive chemicals, including acids and nonaqueous solvents such as those used in GC or HPLC. Filter media is hydrophobic PTFE with a polypropylene housing. Filters have a female Luer-Lok® inlet and a male Luer slip outlet.

Packaging: Bulk packed in zip closure bags.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR