Nalgene® Syringe Filters, Surfactant-Free Cellulose Acetate and Cellulose Acetate, 25mm, Thermo Scientific
Supplier: Thermo Fisher Scientific
723-2520 723-2545 723-2580 723-9920 723-9945 724-2020 724-2045 724-2080
10218-470CS 442.22 CAD
10218-470 10218-472 10218-474 10218-476 10218-478 10218-480 10218-482 10218-484
Nalgene® Syringe Filters, Surfactant-Free Cellulose Acetate and Cellulose Acetate, 25mm, Thermo Scientific
Filters Syringe Filters
Filter sample volumes of 10 to 100 ml with these 25 mm Ø syringe filters.
  • Sterile filters are certified sterile, non-cytotoxic and non-pyrogenic
  • SFCA syringe filters have a blue housing ring
CA membranes have some protein binding and extractables, making them more suitable for clarification of buffers, aqueous solutions and reagents. CA syringe filters have a red housing ring. All housings are polypropylene with female Luer-Lok®inlet and male Luer slip outlet.
SFCA membranes are low protein binding and have lower extractables than standard CA soMore Product Information