Aquastar™ Water Standards for Karl Fischer Titration, MilliporeSigma
Supplier: MilliporeSigma
1.06664.0100 1.12939.0010 1.88054.0005 1.88055.0010 1.88052.0012 1.09259.0250
CA1.06664.0100EA 320.49 CAD
CA1.06664.0100 CA1.12939.0010 CA1.88054.0005 CA1.88055.0010 CA188052.0010 CA1.09259.0250
Aquastar™ Water Standards for Karl Fischer Titration, MilliporeSigma
Standards Water Determination (by Karl Fischer method) Standards
The Aquastar™ range of standards for Karl Fischer titration comprises liquids and solids with defined water contents for a wide variety of applications.
  • Liquids and solids with defined water contents for a wide variety of applications
  • Water standards for both volumetric and coulometric titration
Aquastar™ standards for Karl Fischer titration can be used for monitoring and qualification of your Karl Fischer instruments, for checking and evaluating measuring results, and for performing titer determination of volumetric Karl Fischer reagents.
Merck have introduced into the range of Supelco® SmartStandards, some packs with additional technology - Supelco® SmartStandards that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tags.