ISO17025 ASTM Certified Liquid-in-Glass Mercury Thermometers, Thermco Products
Supplier: Thermco
ACC001CFC ACC001FFC ACC002CFC ACC002FFC ACC003CFC ACC003FFC ACC004CFC ACC004FFC ACC005CFC ACC005FFC ACC006CFC ACC006FFC ACC009CFC ACC009FFC ACC010CFC ACC010FFC ACC011CFC ACC011FFC ACC014CFC ACC014FFC ACC023CFC ACC024CFC ACC025CFC ACC027CFC ACC031FFC ACC033CFC ACC033FFC ACC034CFC ACC034FFC ACC035CFC ACC035FFC ACC036CFC ACC037CFC ACC038CFC ACC039CFC ACC040CFC ACC041CFC ACC042CFC ACC057CFC ACC057FFC ACC061CFC ACC061FFC ACC071CFC ACC071FFC ACC075FFC ACC082CFC ACC082FFC ACC083CFC ACC083FFC ACC084CFC ACC084FFC ACC085CFC ACC085FFC ACC086CFC ACC086FFC ACC087CFC ACC087FFC ACC088CFC ACC088FFC ACC089CFC ACC090CFC ACC091CFC ACC092CFC ACC093CFC ACC094CFC ACC095CFC ACC096CFC ACC119CFC ACC119FFC ACC133CFC ACC134CFC ACC135CFC ACC135FFC ACC137CFC ACC049CFC ACC053CFC ACC100CFC ACC101CFC ACC102CFC ACC103CFC ACC104CFC ACC105CFC ACC106CFC ACC107CFC
10747-390EA 815.61 CAD
10747-390 10747-392 10747-394 10747-396 10747-398 10747-400 10747-402 10747-404 10747-406 10747-408 10747-410 10747-412 10747-422 10747-424 10747-426 10747-428 10747-430 10747-432 10747-440 10747-442 10747-476 10747-478 10747-480 10747-484 10747-496 10747-498 10747-500 10747-502 10747-504 10747-506 10747-508 10747-510 10747-512 10747-514 10747-516 10747-518 10747-520 10747-522 10747-548 10747-550 10747-564 10747-566 10747-604 10747-606 10747-620 10747-634 10747-636 10747-638 10747-640 10747-642 10747-644 10747-646 10747-648 10747-650 10747-652 10747-654 10747-656 10747-658 10747-660 10747-662 10747-664 10747-666 10747-668 10747-670 10747-672 10747-674 10747-676 10747-710 10747-712 10747-746 10747-748 10747-750 10747-752 10747-758 10747-836 10747-844 10747-858 10747-860 10747-862 10747-864 10747-866 10747-868 10747-870 10747-872
ISO17025 ASTM Certified Liquid-in-Glass Mercury Thermometers, Thermco Products
Thermometers Analog Thermometers
These high precision liquid-in-glass mercury thermometers are hand blown, which bear graduations, numbers and letters etched into the glass and filled with and inert gas above the mercury column
ASTM thermometers are used for a variety of specified tests which quantify and qualify repeatable test results in the fields of oil & petroleum testing.
WARNING—Mercury has been designated by EPA and many state agencies as a hazardous material that can cause central nervous system, kidney and liver damage. Mercury, or its vapor, may be hazardous to health and corrosive to materials. CautionMore Product Information