1,3-Bis(diphenylphosphino)propanenickel(II) chloride ≥98.0%

Supplier: TCI America
Warning

Synonyms: [1,3-Bis(diphenylphosphino)propane]nickel (II) chloride, Dichloro[bis(1,3-diphenylphosphino)propane]nickel(II), 1,3-Bis(diphenylphosphino)propane nickel(II)chloride, Dichloro[1,3-bis(diphenylphosphino)propane]nickel(II), [1,3-Bis(diphenylphosphino)propane]dichloronickel(II), [1,3-Bis(diphenylphosphino)propane]nickel(II) dichloride

B1313-25G B1313-5G
TCB1313-25GEA 173.8 CAD
TCB1313-25G TCB1313-5G
1,3-Bis(diphenylphosphino)propanenickel(II) chloride ≥98.0%
1,3-Bis(diphenylphosphino)propanenickel(II) chloride

CAS Number: 15629-92-2
MDL Number: MFCD00015318
Molecular Formula: C27H26Cl2NiP2
Molecular Weight: 542.05
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 213


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Activation Reaction of Unactivated Bonds, Cross-coupling Reaction using Transition Metal Catalysts, Materials Chemistry, Precise Polymerization Reagents, Polymerization Catalysts
Other Product Names / Synonyms: [1,3-Bis(diphenylphosphino)propane]dichloronickel(II), Dichloro[1,3-bis(diphenylphosphino)propane]nickel(II)

Formula: C₂₇H₂₆Cl₂NiP₂
MW: 542.042 g/mol
Melting Pt: 213 °C
Storage Temperature: Ambient
MDL Number: MFCD00015318
CAS Number: 15629-92-2
EINECS: 229-791-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf5e6d0030e04a11928fefb33b8dc724443d904e&vid=cf5e6d0030e04a11928fefb33b8dc724443d904e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR