Seamless Stainless Steel Beakers, Medegen
Supplier: Medegen Medical Products
79310 78730 79310 78730 78740 78720
36311-312CS 721.57 CAD
36311-312 36311-318 36311-319 36311-314 36311-315 36311-316
Seamless Stainless Steel Beakers, Medegen
Beakers
Type 304 stainless steel beakers feature a seamless construction for easy cleaning
Beakers are non-graduated and feature straight sides