Crumble Disk Treats, Bio-Serv
Supplier: Bio-Serv
F05048CD F05048CD F05048CD-25 F05048CD-25
CA89067-296PK 527.66 CAD
CA89067-296 CA89067-298 89067-298 CA89408-188 76325-042 CA76325-042
Crumble Disk Treats, Bio-Serv
Animal Supplies Animal Feed
Compressed mixed fruit–flavored foraging crumbles are a nutritionally complete treat with Vitamin C.
  • Treats provide natural foraging and picking behaviors
  • Can be used with several Bio-Serv devices
  • Uncolored Banana Flavor is Certified (Contaminant Screened) and suitable for use in GLP and TOX studies
  • Nutritionally assayed
  • Shelf Life: 6 months, cool dry storage
Ingredients: Dextrose, Casein, Cellulose, Corn Starch, Sucrose, Mineral Mix, Canola Oil, Maltodextrin, Tablet Binder, Choline Dihydrogen Citrate, Vitamin Mix, Ascorbic Acid, DL-Methionine, BHA,More Product Information