BD Integra™ Syringes with Needles, BD
Supplier: BD
305270 305271
CABD-305269CS 274.94 CAD
CABD-305269 CABD-305270 CABD305271 CABD305272 CABD305273 CABD305274 CABD305283
BD Integra™ Syringes with Needles, BD
Syringes Syringes, Medical
This retracting device delivers complete medication whether activated before or after the needle is withdrawn from the patient.
  • Sterile, disposable
  • Uses only 0.026 ml of waste space
Uses detachable needles with BD PrecisionGlide* technology for patient comfort.