Water nuclease-free, DEPC treated
Supplier: GROWCELLS
Synonyms:
Hydrogen oxide
UPW-0125
UPW-0500
UPW-1000
UPW-2000
UPW-010L
UPW-010Q
10220-380CS
2497.19
CAD
10220-380
10220-382
10220-384
10220-386
10220-388
10220-390
Water nuclease-free, DEPC treated
Water
Sterile molecular biology grade nuclease free water. Prepared with 18.2 megOhm water, 0.001% DEPC treated overnight, 0.1 micron sterile filtered and autoclaved in final packaging.
Water and buffers in IV bags are not injectable. They are not a medical device and are not to be used for intravenous use. They are for research only.
Formula:
H₂O Boiling Pt: 100 °C (1013 hPa) Melting Pt: 0 °C Density: 1 g/cm³ (4 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 EINECS: 231-791-2 Merck Index: 13,10098 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...