Water for PCR nuclease-free

Supplier: GROWCELLS

Synonyms: Hydrogen oxide

PCPW-1000 PCPW-0500 PCPW-0050
10220-392CS 29321.66 CAD
10220-392 10220-394 10220-396
Water for PCR nuclease-free
Water

Sterile PCR grade nuclease free water. Prepared with 18.2 megOhm, 0.1 micron sterile filtered water and autoclaved.

Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR