Water for molecular biology nuclease-free
Supplier: GROWCELLS
NUPW-0050
NUPW-0125
NUPW-0500
NUPW-1000
NUPW-2000
NUPW-010L
NUPW-010LQ
NUPW-020L-Q
10220-398CS
2892.9
CAD
10220-398
10220-400
10220-402
10220-404
10220-406
10220-408
10220-410
10220-412
Water for molecular biology nuclease-free
Water
Sterile molecular biology grade nuclease free water. Prepared with 18.2 megOhm, 0.1 micron sterile filtered water and autoclaved in final packaging.
Formula:
H₂O Boiling Pt: 100 °C (1013 hPa) Melting Pt: 0 °C Density: 1 g/cm³ (4 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 EINECS: 231-791-2 Merck Index: 13,10098 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...