KIMTECH PURE® A7 Cleanroom Lab Coat, Kimberly-Clark
Supplier: Microflex
47655 47657 47652 47653 47654
CA10672-492CS 212.3 CAD
CA10672-492 CA10672-494 CA10672-496 CA10672-510 CA10672-512 CA10672-514
KIMTECH PURE® A7 Cleanroom Lab Coat, Kimberly-Clark
Disposable Garments
Looking for increased protection and superior contamination control? Cover up with the KIMTECH PURE® A7 Cleanroom Lab Coat
  • Enhanced chemical and biological protection
  • Thumb loops to maintain glove and garment interface
  • Low lint to reduce contamination risk
  • Advanced Barrier Fabric for better liquid and dry particle protection
  • High mandarin collar reduces the risk of contamination
  • Elastic wrist and thumb loops for full wrist coverage
  • Recyclable through the RightCycle™ by Kimberly-Clark Professional®
  • Bulk cleanroom packaging
It all adds up to a safer,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Waste Reducing
Gender
Garment application Cleanroom Cleanroom