NIST Traceable pH Buffers with Temperature Table
Supplier: METTLER TOLEDO(INDUSTRIAL) CA
63056121 63056125 63056129 63056116 63056118 63056126 63056122
CA97007-784EA 48.52 CAD
CA97007-784 CA97007-786 CA97007-788 CA97007-790 97007-794 97007-806 97007-800
NIST Traceable pH Buffers with Temperature Table
Buffers pH Calibration Buffers
NIST-traceable buffer for pH electrode calibration and verification.
  • Temperature table on bottle
Measurement results depend on accurate calibrations, and accurate calibrations come from high quality solutions. Each standard solution comes with a certificate of analysis, providing traceability to NIST standards.