Eppendorf®, Vis Cuvettes
Supplier: Eppendorf
0030079353 0030079345 0030079353 0030079345
89498-088EA 521.98 CAD
89498-088 89498-090 CA89498-088 CA89498-090
Eppendorf®, Vis Cuvettes
Cuvettes Spectrophotometer Cuvettes
The Eppendorf Vis Cuvettes are disposable cuvettes made of clear plastic with a light transmission of 300 nm to 900 nm
The Vis cuvettes are the perfect tool for applications outside of the UV range, for example, colorimetric protein assays (Bradford, Lowry, etc.), determining of the optical density of bacterial cultures (OD600 methods), and kinetic and fluorescence measurements. Two diff erent cuvette sizes (semi-micro and macro) for a wide variety of volumes.