Cultubes™, Plastic Culture Tubes with Dual Position Caps, Sterile, Simport Scientific
Supplier: Simport Scientific
T415-2 T415-6 T415-2A T415-6A T416-3 T416-6 T416-6A T405-6 T405-1 T415-3 T416-2A T415-2A T415-3 T416-2A T416-3
CA48648-650CS 354.14 CAD
CA48648-650 CA48648-654 CA48648-656 CA48648-658 CA48648-662 CA48648-664 CA48648-668 CA60831-000 CA73960-188 CA73960-192 CA73960-194 CA73960-202 CA73960-204 CA73960-208 CA73960-434 CA48648-652 CA48648-666 CA60830-196 CA60830-198 CA60831-005 CA73960-190 CA73960-196 CA73960-201 CA73960-206 490010-456 490010-458 490010-460 490010-462
Cultubes™, Plastic Culture Tubes with Dual Position Caps, Sterile, Simport Scientific
Tubes Culture Tubes
These disposable, sterile tubes can be used for most routine laboratory procedures.
  • Pyrogen-free
Transparent polystyrene tubes will withstand moderate centrifugation speeds (1400 g) and temperatures to 70 °C (158 °F). Translucent polypropylene tubes can be centrifuged at higher speeds (3000 g) and resist temperatures from –190 to 120 °C (–310 to 248 °F).
Tubes are supplied with either a 2-position ribbed polyethylene cap (which can be left loose for aerobic work or sealed for anaerobic cultures), or without caps for general-purpose work.
Tubes are placed in a convenient space saving plastic tray oriented horizontally. State-of-the-art packaging keeps tubes neatly aligned for easy manipulation. For users who prefer more compact packaging, see T405/T406 series.
They are biologically inert and exempt from mold release agents. Precision molding with virgin thermoplastics ensures that the tubes will be uniform in size and shape. High resistance to breakage reduces danger in handling infectious or other potentially harmful cultures.
Ordering information: All tubes are supplied sterile.