Motor DC Solar Miniature 1040RPM SHA
Supplier: Solar Made
MC-05/07 MC-05/07
470135-764EA 25.8 CAD
470135-764 470094-400
Motor DC Solar Miniature 1040RPM SHA
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Operates 1/2 to 3 volts, 100ma start-up / runs at 15ma no load.  Strong torque - 300 RPM @ 1/2 volt. Great for gear drives.  2mm shaft dia. Size 1 1/4" dia x 1 1/4" long. 