Bochem Anodized Lab Support Jacks with Stainless Steel Lifts, BrandTech
Supplier: Brandtech
B11016 B11021 B11031 B11041 B11081 B11091
12000-758EA 697.19 CAD
12000-758 12000-760 12000-762 12000-764 12000-766 12000-768
Bochem Anodized Lab Support Jacks with Stainless Steel Lifts, BrandTech
Laboratory Jacks
Improved–Jacks now feature thicker scissor mechanism supports for enhanced stability.
  • 304 stainless lift mechanisms for long service life
  • Anodized aluminum decks
Lift mechanism is constructed entirely of 304 stainless steel with POM rollers and phenolic knob for long service life in corrosive environments, such as fume hoods. All-roller design provides smooth stable height adjustment. Large (30x30cm and 40x40cm) sizes are supplied with a ratcheting wrench for easy height adjustment. Jacks are manufactured according to DIN 12897.