SiliaPrep™ Silica and Functionalized Silica SPE Cartridges, SiliCycle
Supplier: SiliCycle
ER31030B06P ER32030B01B ER32030B01C ER32030B03G ER32030B03P ER32030B06P ER32030B06S ER32030B06U ER32030B12U ER33030B01B ER33030B01C ER33030B03G ER33030B03P ER33030B06P ER33030B06S ER33030B06U ER33030B12U ER63730B01C MR023830B01C MR023830B06S MR023830B12U MR028030B01C MR028030B06S MR028030B12U MR802830B01C MR802830B06S MR802830B12U ER31030B06P ER32030B01B ER32030B01C ER32030B03G ER32030B03P ER32030B06P ER32030B06S ER32030B06U ER32030B12U ER33030B01B ER33030B01C ER33030B03G ER33030B03P ER33030B06P ER33030B06S ER33030B06U ER33030B12U ER63730B01C MR023830B01C MR023830B06S MR023830B12U MR028030B01C MR028030B06S MR028030B12U MR802830B01C MR802830B06S MR802830B12U
75958-254BX 544.04 CAD
75958-254 75958-306 75958-308 75958-310 75958-312 75958-314 75958-316 75958-318 75958-320 75958-322 75958-324 75958-326 75958-328 75958-330 75958-332 75958-334 75958-336 75958-718 75959-374 75959-382 75959-386 75959-422 75959-430 75959-434 75959-458 75959-466 75959-470 CA75958-254 CA75958-306 CA75958-308 CA75958-310 CA75958-312 CA75958-314 CA75958-316 CA75958-318 CA75958-320 CA75958-322 CA75958-324 CA75958-326 CA75958-328 CA75958-330 CA75958-332 CA75958-334 CA75958-336 CA75958-718 CA75959-374 CA75959-382 CA75959-386 CA75959-422 CA75959-430 CA75959-434 CA75959-458 CA75959-466 CA75959-470
SiliaPrep™ Silica and Functionalized Silica SPE Cartridges, SiliCycle
Chromatography Columns
Optimize your SPE protocols by using SiliCycle SiliaPrep™ SPE Cartridges.
  • High quality and wide variety of SiliaBond sorbents available
  • Very good separation (very tight particle size distribution and no fines)
  • High recoveries and yields
  • Less time and solvent spent conditioning the sorbent
  • No silica contamination of your final product
  • Reproducible flow rate from lot-to-lot
  • Excellent packing and storage qualities
SiliaPrep are available in different formats; cartridges (SiliaPrep SPE Cartridges), and well plates (SiliaPrep 96 Well Plates) with various sorbents (SiliaFlash® and SiliaBond®), and bed weight (up to 20 grams). The well plates are used in high throughput combinatorial chemistry, drug discovery and screening, and metabolism pharmacokinetics applications and for automated method as multiprobe approach.