ActivArmr® 97-011 High-Visibility Nitrile Gloves, 3/4 Dipped, Ansell
Supplier: Ansell Healthcare
256064 256066 256067
10510-826CS 2010.2 CAD
10510-826 10510-828 10510-830 10510-832
ActivArmr® 97-011 High-Visibility Nitrile Gloves, 3/4 Dipped, Ansell
Gloves
Protecting workers from the elements, ActivArmr® 97-011 gloves deliver incomparable high-visibility hand protection for cold, wet environments.
  • Superior cold protection
  • Excellent handling of wet, dry, and oily materials
  • High-visibility gloves help avoid injuries
Colder temperatures impair reflex time, dexterity, grip and feel.
Visibility is a top priority in many work environments. It can help avoid injuries and save lives. ActivArmr® 97-011 Cold Weather Hi-Viz gloves are specially designed with an exclusive black nitrile coating that contrastsMore Product Information