TouchNTuff® 93-300 Nitrile Gloves with Long Cuff, Powder-Free, Ansell
Supplier: Ansell Canada
556079 556080 556081 556082
10049-908CS 756.87 CAD
10049-908 10049-910 10049-913 10049-915
TouchNTuff® 93-300 Nitrile Gloves with Long Cuff, Powder-Free, Ansell
Gloves
These gloves offer three times the puncture resistance of latex disposables with comparable thickness and deliver a high level of comfort and increased protection in the forearm with a longer cuff.
  • Gloves are packed in Class 100 Cleanroom environment
  • Cleanroom compatible packaging with IPA-resistant ink
  • Recommended for Class 100 (ISO 5)
  • Grade A Cleanroom suitable
  • High degree of dexterity for precision work
  • Silicone-free
  • High level of comfort and increased forearm protection
The thin structure ensures the level of dexterity required for precision work.
More Product Information