Mixing Bulb Style, Wide-Mouth Volumetric Flasks, Class A, Heavy Wall with [ST] Stoppers, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
623020-0100 623020-0200 623020-0250 623020-0300 623020-0400 623020-0500 623020-1000
KT623020-0100EA 264.78 CAD
KT623020-0100 KT623020-0200 KT623020-0250 KT623020-0300 KT623020-0400 KT623020-0500 KT623020-1000
Mixing Bulb Style, Wide-Mouth Volumetric Flasks, Class A, Heavy Wall with [ST] Stoppers, Kimble Chase, DWK Life Sciences
Flasks Volumetric Flasks
Wide mouths allow easy access into these mixing bulb volumetric flasks for applications such as dissolving tablets and other solids
Designed from ASTM Specification E288, Class A requirements . Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Heavy wall design provides added durability. Mixing bulb in neck accommodates approximately one tenth of the flask’s total volume and is designed to aid in dissolving or mixing the contents of the flask. Calibrated to contain, with graduation ring blasted on the neck
Ordering information: Standard Taper ground glass pennyhead stopper included.