GsBP®-5 Non-Polar GC Columns, GS-Tek
Supplier: General Separation Technologies, Inc.
0510-1010 0510-1017 0510-2010 0510-2040 0518-1001 0518-1004 0518-1501 0518-2001 0518-2003 0518-2004 0518-4001 0520-1203 0520-1502 0520-1703 0520-2501 0520-5005 0525-0501 0525-1501 0525-1501HT 0525-1502 0525-1505 0525-1510 0525-2502 0525-3001 0525-3001HT 0525-3002 0525-3005 0525-3010 0525-6010 0532-1001HT 0532-1005 0532-1501 0532-1501HT 0532-1502 0532-1505 0532-1510 0532-2502 0532-2505 0532-2510 0532-3001 0532-3001HT 0532-3002 0532-3005 0532-3010 0532-3015 0532-5001 0532-5002 0532-5005 0532-5010 0532-6001 0532-6002 0532-6005 0532-6010 0532-6030 0532-6050 0553-1026 0553-1050 0553-1502 0553-1505 0553-1510 0553-1515 0553-1530 0553-1550 0553-2550 0553-3002 0553-3005 0553-3008 0525-5002 0525-6001 0525-6002 0525-6005 0553-3010 0553-3015 0553-3026 0553-3030 0553-3050 0553-6015 0553-6030 0520-2503 0520-2505 0520-5001 0520-5003 0553-6050
10498-874EA 998.56 CAD
10498-874 10498-876 10498-882 10498-884 10498-886 10498-888 10498-890 10498-892 10498-894 10498-896 10498-898 10498-900 10498-902 10498-904 10498-906 10498-908 10498-910 10498-912 10498-914 10498-916 10498-918 10498-920 10498-922 10498-924 10498-926 10498-928 10498-930 10498-932 10498-934 10498-936 10498-938 10498-940 10498-942 10498-944 10498-946 10498-948 10498-952 10498-954 10498-956 10498-958 10498-960 10498-962 10498-964 10498-966 10498-968 10498-970 10498-972 10498-974 10498-976 10498-978 10498-980 10498-982 10498-984 10498-986 10498-988 10498-990 10498-992 10498-994 10498-996 10498-998 10499-000 10499-002 10499-004 10499-006 10499-008 10499-010 10499-012 10499-054 10499-056 10499-058 10499-060 10499-062 10499-064 10499-066 10499-068 10499-070 10499-072 10499-074 10499-300 10499-302 10499-304 10499-306 10499-316
GsBP®-5 Non-Polar GC Columns, GS-Tek
Chromatography Columns
Amines & Phenols, Amphetamines and Precursors - TMS Derivative, Antihistamines, Aroclors 1016-1268 (without 1221), Bacterial Fatty Acid Methyl Esters, Basic Drugs (Underivatized), Benzodiazepines (Basic Drugs - Underivatized), Bergamot Oil, Butter Triglycerides, Cannabinoids (TMS Derivatives), Chlorinated hydrocarbons, CLP Pesticides, Cocaine & Metabolites (TMS Derivatives), Cold Medication, Cold-Pressed Orange Oil, Common Drug Screen, Congeners in DIN Method PCBs, Ethanolamines, Typical Application: Ethyleneamines, European Red List Volatiles, Garlic Oil, Herbicides, Lemon Oil, Marijuana - TMS Derivatives, Major Metabolites - TMS Derivatives, Motor Oil, Narcotics and Adulterants, Opiates (TMS Derivatives), Organic Volatile Impurities, Organochlorine Pesticides, Organomercury Compounds, Organotin Compounds II, Pain Killers - TMS Derivatives, PBDEs, PCBs by EPA Method 8083, Phencyclidine (PCP), Phytosterols (Saw Palmetto), Steroids: Anabolic (Underivatized), Sympathomimetic Amines Derivatives, Ylang Ylang Oil.
  • 5% phenyl 95% methyl polysiloxane phase
  • Equivalent to USP G27, G41, G66
Ordering information: These columns are suitable replacements for HP-5, DB-5, CP-Sil 8CB, VF-5MS, Rtx-5, Rxi-5, SP-, ZB-5, BP-5.