GsBP®-1 Non-Polar GC Columns, 5 - 25 m, GS-Tek
Supplier: General Separation Technologies, Inc.
0120-1203 0120-1701 0110-2010 0115-1012 0118-1001 0118-1002 0118-1004 0118-2001 0118-2004 0125-1502 0125-1510 0125-2501 0125-2502 0132-1550 0132-2501 0132-2502 0132-2505 0132-2510 0153-0501HT 0153-0508 0153-0526 0153-0550 0153-0715 0153-0750 0153-1008 0153-1026 0153-1050 0153-1501 0153-1502 0153-1505 0153-1510 0153-1515 0153-1530 0153-1550 0153-2510 0153-2550 0153-A550 0120-2503 0120-2505 0125-1501 0125-1501HT 0132-1501 0132-1501HT 0132-1502 0132-1510 0120-2501 0110-0512 0110-1010 0132-1530
10498-084EA 753.06 CAD
10498-084 10498-086 10498-676 10498-682 10498-684 10498-686 10498-688 10498-690 10498-692 10498-694 10498-696 10498-698 10498-700 10498-718 10498-720 10498-722 10498-724 10498-726 10498-760 10498-762 10498-764 10498-766 10498-768 10498-770 10498-772 10498-774 10498-776 10498-778 10498-780 10498-782 10498-784 10498-786 10498-788 10498-790 10498-792 10498-794 10498-832 10499-014 10499-016 10499-026 10499-028 10499-036 10499-038 10499-040 10499-042 10499-292 10499-294 10499-296 10499-308
GsBP®-1 Non-Polar GC Columns, 5 - 25 m, GS-Tek
Chromatography Columns
GsBP™ GC columns with high quality column performances in all aspects.
  • 100% poly(methylsiloxane) phase
  • Equivalent to USP G1, G2, G38
Typical Application: Aldehyde and Ketone, Anabolic Steriods, Anticonvulsants, Aromatics I, Aromatics in Gasoline - ASTM Method 5769, Citronella Java Oil, Common Industrial Solvents, Denatured Fuel Ethanol - ASTM D5501, Diesel Fuel, DNPH Derivative, Drug Screen, EPA Method 551, EPA TO-14 Compounds, Esters I, Ethylene Glycol Mixture, Eucalyptus Oil, Camphor, Menthol, Fatty Acids (Free), Flavor Volatiles, FragranceMore Product Information