GsBP®-Tubing Fused Silica GC Column, GS-Tek
Supplier: General Separation Technologies, Inc.
9010-0500 9010-1000 9018-0500 9018-1000 9020-0500 9020-1000 9020-1500 9020-5000 9025-0500 9025-1000 9025-1500 9032-0500 9032-1000 9032-1500 9032-5000 9053-0500 9053-1000 9053-1500
10500-072EA 436.16 CAD
10500-072 10500-074 10500-076 10500-078 10500-080 10500-082 10500-084 10500-086 10500-088 10500-090 10500-092 10500-094 10500-096 10500-098 10500-100 10500-102 10500-104 10500-106
GsBP®-Tubing Fused Silica GC Column, GS-Tek
Chromatography Columns
GC tubing.
Order Now
SPECIFICATIONS
Environmentally Preferable
Temperature limit -60 to 325/350°C -60 to 325°C