GsBP®-PLOT-Q GC Columns, GS-Tek
Supplier: General Separation Technologies, Inc.
8653-2530 8653-3030 8653-3030PT 8653-5030 8625-3008 8632-1515 8632-1515PT 8632-3015 8653-1030 8653-1530 8653-1530PT
10499-716EA 1667.8 CAD
10499-716 10499-718 10499-720 10499-722 10500-052 10500-054 10500-056 10500-058 10500-062 10500-064 10500-066
GsBP®-PLOT-Q GC Columns, GS-Tek
Chromatography Columns
Typical Application: Air, Carbon Monoxide, Methane, Carbon Dioxide, Alcohols, Fatty Acids (Free), Gases (Air, Carbon Monoxide, Methane, Carbon Dioxide), Hydrocarbon Gases, N2OI, Natural Gas, Nitrogen, Oxygen, Argon, Oxygenates, Refinery Gas I, Solvents, Sulfur Gas Analysis in Light HydroCarbon Streams.
  • Divinylbenzene porous polymer
  • Immobilized
  • Equivalent to USP S2, S3, S6
Ordering information: These columns are suitable replacements for HP-PLOT Q, CP-Porapak Q, CP-Porabond Q,Rt-Q.