GsBP®-GasPro GC Columns, GS-Tek
Supplier: General Separation Technologies, Inc.
8532-0505 8532-1505 8532-3005
10500-046EA 953.55 CAD
10500-046 10500-048 10500-050
GsBP®-GasPro GC Columns, GS-Tek
Chromatography Columns
Typical Application: C1 and C2 halocarbons (freons), extended hydrocarbon analysis, halocarbons, halothane, inorganic gases, ,ercaptans, residue solvents, sulfur compounds in propylene (1 ppm), sulfur gas analysis in light hydrocarbon streams I.
  • PLOT phase
  • Immobilized
Ordering information: These columns are suitable replacements for GS-GasPro, CP-Select Silica.