GsBP®-PLOT-MS5A GC Columns, GS-Tek
Supplier: General Separation Technologies, Inc.
8432-1512 8432-3012 8453-1525 8453-1550 8453-3025 8453-3050
10500-032EA 1553.8 CAD
10500-032 10500-034 10500-038 10500-040 10500-042 10500-044
GsBP®-PLOT-MS5A GC Columns, GS-Tek
Chromatography Columns
Typical Application: Air, Argon, CO, Hydrogen, Natural gas, Nitrogen Oxides, Noble gases, Refinery has, SF6.
  • PLOT phase
  • Molecular sieve 5A
  • Immobilized
Ordering information: These columns are suitable replacements for HP-PLOT Molsieve, CP-PLOT Molesieve.