GsBP®-PLOT-S GC Columns, GS-Tek
Supplier: General Separation Technologies, Inc.
8225-2505 8225-3005 8232-1508 8232-3008 8232-5008 8253-1515 8253-3015 8253-5015
10500-000EA 1751.64 CAD
10500-000 10500-002 10500-004 10500-006 10500-008 10500-010 10500-012 10500-014
GsBP®-PLOT-S GC Columns, GS-Tek
Chromatography Columns
Typical Application: Refineral gas, Propane, Propylene, Butadienes, CFCs, C1 to C5, Extended C1 toC10, Nitrogen oxides.
  • PLOT phase
  • Al₃O₃
  • Modified by Na₂SO₄
  • Immobilized
Ordering information: These columns are suitable replacements for HP-PLOT AL₂O₃, CPAlumina PLOT, Rt-Alumina, GS-Alumina.