GsBP®-VMS Mid-Polar GC Columns, GS-Tek
Supplier: General Separation Technologies, Inc.
6418-2010 6425-3014 6425-6014
10499-946EA 1346.49 CAD
10499-946 10499-948 10499-950
GsBP®-VMS Mid-Polar GC Columns, GS-Tek
Chromatography Columns
Equivalent to: VF-624MS, Rtx-VMS.
  • Proprietary phase
  • Close to GsBP-1301
  • Mid-polar
  • USP Code: G43
GC is a common tool for quality control in pharmaceutical production, small molecule research in medical chemistry and drug discovery, metabolism research and other areas. The complexity of these applications demands special columns to conduct separation with a high degree of column inertness and low column bleed for advanced GC, HSGC, HS-GC-MSD and other instrumentation.
Order Now
SPECIFICATIONS
Description 10499-946 10499-948 10499-950
Environmentally Preferable
Temperature limit -20 to 260/280°C -20 to 260/280°C -20 to 260/280°C