GsBP®-624 Mid-Polar GC Columns, GS-Tek
Supplier: General Separation Technologies, Inc.
6218-2010 6218-2010UI 6218-4010 6220-2511 6220-3011 6220-6011 6225-3014 6225-3014UI 6225-6014 6225-6014UI 6232-3018 6232-3018UI 6232-6018 6232-6018UI 6232-9018 6253-1530 6253-3030 6253-3030UI 6253-6030 6253-7530 6253-7530UI 6253-A030 6253-A530 6225-3014-6PK 6225-6014-6PK 6232-3018-6PK
10499-878EA 1296.44 CAD
10499-878 10499-880 10499-882 10499-884 10499-886 10499-888 10499-890 10499-892 10499-896 10499-900 10499-902 10499-906 10499-908 10499-910 10499-912 10499-914 10499-916 10499-920 10499-922 10499-924 10499-926 10499-928 10499-930 10499-932 10510-868 10510-870 10510-872
GsBP®-624 Mid-Polar GC Columns, GS-Tek
Typical Application: Air, water-drinking, water-underground, water-waste, Packaging, Cosmetics, Healthcare, Drinking, Tobacco, Nutrition, Alcohols, FAME, Halogenated Hydeocarbons, Gasoline, Nitrogen Based Solvents II, Residual Solvents, USP 467, Volatile Amines, Volatile Petroleum Hydrocarbons (VPH), Volatile Sulfurs, Alcohols, DMA, DMSO, Esters, Ethers, Glycols II, EPA 501.3, EPA 503.1, EPA 504, EPA 524.2, EPA 601, EPA 602, EPA 624, EPA 8010, EPA 8015, EPA 8020, EPA 8021, EPA 8240, EPA 8260.
  • Proprietary phase
  • Close to GsBP-1301
  • Mid-polar
  • Equivalent to USP G43
Ordering information: These columns are suitable replacements for DB-624, DB-VRX, Rtx-624, Rtx-VMS, DB 502.2, VOCCol.