GsBP®-608 Mid-Polar GC Columns, GS-Tek
Supplier: General Separation Technologies, Inc.
5253-3008 5225-3002 5232-3005 5253-3005
10499-222EA 1782.19 CAD
10499-222 10499-776 10499-778 10499-780
GsBP®-608 Mid-Polar GC Columns, GS-Tek
Chromatography Columns
Similar phases include DB-608.
Ordering information: These columns are suitable replacements for DB-608.
Order Now
SPECIFICATIONS
Description 10499-222 10499-776 10499-778 10499-780
Environmentally Preferable
Temperature limit 40 to 280/300°C 40 to 320/340°C 40 to 320/340°C 40 to 280/300°C