GsBP®-50+MS Mid-Polar GC Columns, GS-Tek
Supplier: General Separation Technologies, Inc.
5032-1501HT 5032-1502 5032-1505 5032-3001 5018-2001 5018-2003 5020-1203 5025-0501 5025-0501HT 5025-1501 5025-1501HT 5025-1502 5025-1505 5025-3001 5025-3001HT 5025-3002 5025-3005 5032-1501 5032-3001HT 5032-3002 5032-3005 5053-0520 5053-1020 5053-3002 5053-3005 5053-3010 5053-3015 5053-1502 5053-1505 5053-1510 5053-1515
10499-214EA 1158.17 CAD
10499-214 10499-216 10499-218 10499-220 10499-654 10499-656 10499-658 10499-660 10499-662 10499-664 10499-666 10499-668 10499-670 10499-672 10499-674 10499-676 10499-680 10499-682 10499-684 10499-686 10499-688 10499-690 10499-694 10499-696 10499-698 10499-700 10499-702 10499-704 10499-706 10499-708 10499-710 10499-712 10499-714 10499-772
GsBP®-50+MS Mid-Polar GC Columns, GS-Tek
Chromatography Columns
Typical Application: Antidepressant and anticonvulsant drug, BHA and BHT, Butter Triglycerides, EPA 625, Phenols, EPA 8081 Organochlorine Pesticides, Free Steroids, Hallucinogens, PCBs, PHAs, Phenols, Tocopherols, Triazine Herbicides II.
  • 50% phenyl 50% methyl polysiloxane phase
  • Mid-polar
  • Equivalent to USP G3
Ordering information: These columns are suitable replacements for DB-17, DB-17MS, HP-50+, CP-Sil 24CB, VF-17MS, Rxt-50, Rtx-17.