GsBP®-35MS Mid-Polar GC Columns, GS-Tek
Supplier: General Separation Technologies, Inc.
3518-2001 3518-2001UI 3520-1503 3520-2503 3525-1502 3525-1502UI 3525-3001 3525-3002 3525-3002UI 3532-1502 3532-1502UI 3532-3002 3532-3002UI 3532-3005 3553-1510 3553-3005 3553-3010
10499-606EA 1463.16 CAD
10499-606 10499-608 10499-610 10499-612 10499-614 10499-616 10499-618 10499-620 10499-624 10499-626 10499-628 10499-630 10499-632 10499-634 10499-636 10499-638 10499-640 10499-642 10499-644 10499-646
GsBP®-35MS Mid-Polar GC Columns, GS-Tek
Chromatography Columns
Typical application: Amytriptyline and Nortriptyline, Anilines, Barbiturates (Acid-neutral drugs), Basic drugs (Underivatized), Benzodiazepines, Chlordane, CLP Pesticides, Cold medication (Basic drugs - Underivatized), Endocrine disruptos: Butyl tins (hexyl derivatives), EPA Method 552.2, EPA Method 615-Chlorophenoxyacid herbicides, Ethanolamines, Fentanyl, Organo Tins, Organochlorine pesticides I EPA Method 8081A, Organochlorine pesticides IV, Organophosphorus pesticides II EPA 8141A, PCBs by EPA Method 8082, Pesticides, EPA 508.1, Phenols, Phenoxy acid herbicides - Methyl Derivative, EPA 8151A, Primary amines, Sympathomimetic amines (Basic drugs - Underivatized), Toxaphene.
  • 35% phenyl 65% methyl polysiloxane phase
  • Mid-polar
  • Equivalent to USP G28, G32 and G42
Ordering information: These columns are suitable replacements for DB-35, DB-35MS, HP-35, Rtx-35, ZB-35,VF-35.